Sensorex UVT-LED Uv Transmittance Monitor, Portable
Representative image only.
ITS Mfr # UVT-LED-H-NA – Item # WW-99522-02
Eliminate drift from multiple lamps and sensors
  • Save money and get stable readings with ultra-long life UV-C LED light source
  • Light source: UV-LED
  • Measuring interval: manually controlled
  • Sample method: dip-style submersion
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±1.0%T
  • DescriptionUv Transmittance Monitor, Portable
  • Warranty1 year
1 Year Warranty
Key Features
  • Save money and get stable readings with ultra-long life UV-C LED light source
  • Light source: UV-LED
  • Measuring interval: manually controlled
  • Sample method: dip-style submersion
  • 1 year warranty
More About this Item
Ultraviolet radiation is a cost-effective method for wastewater and water disinfection. Lamp does not require warm-up time, so on/off is instantaneous and measurements are available immediately on the backlit LCD. Portable and battery-operated — perfect to grab sample readings!