Sensorex FC75P Conductivity Probe, K=0.1 with ATC-P1K, 3/4"NPT(M); 20' Cable
Representative image only.
Sensorex Mfr # 971972 – Item # WW-19066-26
Ideal for laboratory or light industrial use
  • Graphite measuring surfaces
  • PT 1000 RTD temperature compensation
  • 3/4" M NPT mounting colar
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Cell ConstantK = 0.1
  • Electrode MaterialGraphite
  • Body MaterialEpoxy
  • Fitting¾\" NPT(M)
  • Cable Length (m)6
  • Cable Length (ft)20
  • DescriptionConductivity Probe, K=0.1 with ATC-P1K, 3/4\"NPT(M); 20' Cable
  • Warranty1 year
1 Year Warranty
Key Features
  • Graphite measuring surfaces
  • PT 1000 RTD temperature compensation
  • 3/4" M NPT mounting colar
  • 1 year warranty
More About this Item
Economical laboratory or light industrial conductivity sensors feature either graphite or platinum ring technology. The durable epoxy body construction provides a rugged, dependable sensor. Suitable for most conductivity meters.