Secondary Hydrocarbon Filters for 30" Self-Monitoring Ductless Fume Hoods
Representative image only.
Mfr # AU-5504 – Item # WW-33713-55
SPECS & DESCRIPTION
Specifications & Description
  • Hood Width (in)30
  • Exterior Width (in)30
  • Filter TypeSecondary Hydrocarbon
  • DescriptionSecondary Hydrocarbon Filters for 30\" Self-Monitoring Ductless Fume Hoods
More About this Item
Protect yourself from hazardous fumes, vapors, odors, particles, and contaminants with one of four types of filters for 30” self-monitoring ductless fume hoods
The activated secondary hydrocarbon filter provides a high degree of absorbency—ideal for organic vapors, odors and fumes, including alcohols, acetone, xylene, and methylene chloride.