Secondary HEPA Filters for 42" Self-Monitoring Ductless Fume Hoods
Representative image only.
Mfr # AU-5520 – Item # WW-33713-86
SPECS & DESCRIPTION
Specifications & Description
  • Hood Width (in)42
  • Exterior Width (in)42
  • Filter TypeSecondary HEPA
  • DescriptionSecondary HEPA Filters for 42\" Self-Monitoring Ductless Fume Hoods
More About this Item
Protect yourself from hazardous fumes, vapors, odors, particles, and contaminants with one of four types of filters for 42” self-monitoring ductless fume hoods
The secondary HEPA filter has an efficiency rating of 99.997% for collecting particles down to 0.3 microns—use for containment of particles such as toxic powders, asbestos, and lead.