SciMatCo SC5040 Polypropylene Acid Storage Cabinet, 50 X 2.5 L bottle capacity

Representative image only.
SciMatCo Mfr # SC5040 – Item # WW-81780-40
Liquid tight to prevent leakage
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPolypropylene
  • Capacity (Liters)125
  • Capacity (Gal)33
  • Number of Shelves1
  • Width (in)48
  • Height (in)40
  • Depth (InchShort)18
  • Width (cm)121.9
  • Height (cm)101.6
  • Depth (cm)45.7
  • ColorWhite
  • ComplianceOSHA, UFC, IFC
  • Shipping Weight (lbs)126
  • DescriptionPolypropylene Acid Storage Cabinet, 50 X 2.5 L bottle capacity
  • Warranty1 Year
1 Year Warranty

Key Features

  • 1 year warranty

More About this Item

These polypropylene acid storage cabinets contain no metal parts— they won’t corrode or rust. Constructed of 3/8" (0.96 cm)-thick white polypropylene, cabinets feature plastic hinges and welded interior seams to prevent leakage. All cabinets include hazard warning label.

Compliance: OSHA, UFC, IFC

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat