Scientific Instruments 240-364 LED Illuminator, Dual head, Combination beam
Representative image only.
Scientific Instruments Mfr # 240-364 – Item # WW-09809-35
High intensity light for your inspection applications
  • Fully posable goosenecks allow for multiple light configurations
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Light intensity19,000 footcandles at 6\"
  • Lamp Color Temperature3200 K
  • Bulb Life50,000 hours
  • Bulb TypeCombination/24º
  • Mounting TypeTable
  • Arm Length (in)11
  • Cord Length5-ft (1.5 m)
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • DescriptionLED Illuminator, Dual head, Combination beam
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Fully posable goosenecks allow for multiple light configurations
  • Limited Lifetime Warranty
More About this Item
Order the optional magnifier for convenient, close-up inspections.