Scienceware F378451000 Mini-Sieve Set, removable phosphor bronze screens
Representative image only.
Scienceware Mfr # F378451000 – Item # WW-59986-40
For handheld use in the field or lab
  • Removable screens allow you to easily exchange the mesh size for different applications
₹47,400.00INR / Set of 1
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene frame, phosphor Bronze mesh
  • Height (in)8 1/2
  • Outside Diameter (in)2.8
  • Height (InchShort)8.5
  • DescriptionMini-Sieve Set, removable phosphor bronze screens
Key Features
  • Removable screens allow you to easily exchange the mesh size for different applications
More About this Item
Polypropylene sieve set includes four screen holders, a receiving pan, a sieve cover, and eight phosphor bronze screens (25, 35, 45, 60, 80, 120, 170, and 230 mesh). Use any combination of screens for quick and easy particle size sorting. The interior of the screen holder measures 2”H x 1-1/8” dia. Overall size with the receiving pan and sieve cover is 6-7/16”H x 2-7/8” dia.