Scienceware F371740000 Spinbar Rare Earth Octagon Magnetic Stir Bar, 2" X 7/8"
Representative image only.
Scienceware Mfr # F371740000 – Item # WW-04775-28
Improves stirring efficiency by reducing frequency of spinouts
  • Ideal for viscous solutions, high-speed stirring and extended stirring applications
  • Autoclavable
  • 1 year warranty
PRODUCT OPTIONS
₹21,600.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (in)0.875
  • Diameter (cm)2.2225
  • DescriptionSpinbar Smco, Stirring Bar, PTFE, 2\" X 7/8\"
  • Warranty1 year
1 Year Warranty
Key Features
  • Ideal for viscous solutions, high-speed stirring and extended stirring applications
  • Autoclavable
  • 1 year warranty
More About this Item
Samarium Cobalt magnet contains PTFE which provides strong coupling with drive magnets to reduce frequency of spinout. Octagon shape with pivot ring creates strong turbulence with minimal clicking and friction.