Scienceware F134830075 Write-On Tape, 3/4"W x 120', Pack of 4
Representative image only.
Scienceware Mfr # F134830075 – Item # WW-08277-45
Accepts pen, pencil, or solvent ink marker
  • Autoclavable
  • 3 month warranty
PRODUCT OPTIONS
₹13,700.00INR / Pkg of 4
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")1440
  • Width (InchShort)3/4
  • ColorWhite
  • Width (in)3/4
  • Length (ft)120
  • Width (cm)1.9
  • Length (m)36.58
  • DescriptionWrite-On Tape, 3/4\"W x 120', Pack of 4
  • Warranty3 month
3 Month Warranty
Key Features
  • Autoclavable
  • 3 month warranty
More About this Item
White autoclavable tape withstands temperatures from –99 to 250°F (–73 to 121°C). Tapes are on a 3" (7.6 cm) diameter core.