Scienceware Aero Dual Eyewash Station Buffered pH Neutral 16oz

Representative image only.
Scienceware Mfr # 248825016 – Item # WW-12345-72
Provides immediate first aid for eye injuries
  • Effectively rinses impurities and irritants from your eyes
  • Neutralizes acids and alkalis that may enter eyes
  • Offers four year shelf life from date of manufacture
  • Meets all FDA requirements including buffered pH solution and preservative as per 21 CFR part 349
  • Covered station keeps bottles clean throughout shelf life
  • Carrying handle makes transport easy for field work
  • Wall mountable—hardware included
  • 3 month warranty

SPECS & DESCRIPTION

Specifications & Description

  • Width (in)3 3/4
  • Depth (in)1/2
  • Height (in)4 1/4
  • ComplianceANSI Z358.1-2009
  • DescriptionDual Eyewash Station Buffered pH Neutral 16oz
  • Warranty3 month
3 Month Warranty

Key Features

  • Effectively rinses impurities and irritants from your eyes
  • Neutralizes acids and alkalis that may enter eyes
  • Offers four year shelf life from date of manufacture
  • Meets all FDA requirements including buffered pH solution and preservative as per 21 CFR part 349
  • Covered station keeps bottles clean throughout shelf life
  • Carrying handle makes transport easy for field work
  • Wall mountable—hardware included
  • 3 month warranty

More About this Item

The Aero Emergency Eye Wash combines sterile saline with buffered, pH solution to provide fast, safe, and effective first aid for eye injuries. The all-in-one solution quickly neutralizes acid and corrosive chemicals, returning eyes to normal 7.4 pH levels. Ideal for use in laboratory, classroom, work site, and public facilities.

Compliance: ANSI Z358.1-2009

Available for sale in the United States only.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat