Scienceware 38000 Hot Hand Rubber Mitt, One Size Fits All; 1/Pk
Representative image only.
Scienceware Mfr # 38000-0000 – Item # WW-06910-00
₹4,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • SizeOne size fits all
  • GripStudded
  • SterileNo
  • DescriptionHot Hand Rubber Mitt, One Size Fits All; 1/Pk
More About this Item
The Hot Hand® silicone rubber mitt protects your hand from hot labware. Gripping surfaces are studded for poistive grip; hole tabs on either end are for hanging. Maximum temperature is 500°F (260°C). Fits any glove size.