Scienceware 37080-0020 Vortex attachment, 1.5 mL, 20 tubes held
Representative image only.
Scienceware Mfr # 37080-0020 – Item # WW-04723-61
₹25,700.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Number of Tubes20
  • Tube Size1.5 mL
  • DescriptionVortex attachment, 1.5 mL, 20 tubes held
More About this Item
Use these vortex attachments to agitate microcentrifuge tubes. Attachments work with any vortex shaker that has a rubber pad or test tube cup. Sliding disc keeps tube caps securely closed during shaking. Unit is constructed of sturdy plastic. Bel-Art.