Scienceware 20197-1515 PTFE Joint Stopper for Standard Taper Size 24/40; 2/Pk
Representative image only.
Scienceware Mfr # 20197-1515 – Item # WW-06133-70
₹27,800.00INR / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • Height (mm)40
  • Taper Size24/40
  • ColorRed-White
  • MaterialPolytetrafluoroethylene (PTFE)
  • DescriptionPTFE Joint Stopper for Standard Taper Size 24/40; 2/Pk
More About this Item
PTFE stoppers have a polypropylene turning nut to eliminate frozen neck joints. Use up to 410°F
(210°C).