This item is now discontinued. We recommend the following as suggested replacements: WW-06136-53
Scienceware 18307-0015 vinyl-covered lead ring weight for 500 to 2000 mL flasks
Representative image only.
Scienceware Mfr # 18307-0015 – Item # WW-06137-04
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Liters)2
  • MaterialVinyl-Covered
  • DescriptionScienceware 18307-0015 vinyl-covered lead ring weight for 500 to 2000 mL flasks
More About this Item
Keep labware stable in baths or anywhere flasks might float, spill, or break. Bright red vinyl covering will not scratch flasks.