Sartorius QP1 Cubis II Pharma Software Package
Representative image only.
Sartorius Mfr # QP1 – Item # WW-39742-56
Optimize performance of your Sartorius balance
  • Choose software packages based on your application needs
PRODUCT OPTIONS
₹401,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCubis II Pharma Software Package
Key Features
  • Choose software packages based on your application needs
More About this Item
Add these software packages for instance, lab applications, functions, or utilities. The licenses for these packages can be ordered together with the balance and pre-installed on the balance and ready for use after delivery.
This software package contains applications concerning the topic compliance with pharmaceutical-relevant guidelines as 21 CFR Part 11 and USP 39, Chapter 41. The package also includes applications such as user management, digital signatures, audit trail, USP minimum weight.