Sartorius Presterilized absorbent pads, 47 mm, 1000/box

Representative image only.
Sartorius Mfr # 15410--47----ALR – Item # WW-81055-42
Ideal for use in culturing microorganisms
  • Tested for absorption of 1.8 to 2.2 mL of Culture media
  • pH neutral
  • Individually packaged to assure sterility
  • 3 month warranty

SPECS & DESCRIPTION

Specifications & Description

  • Outside Diameter (OD) (in)1 7/8
  • Outside Diameter (OD) (cm)4.7
  • DescriptionPresterilized absorbent pads, 47 mm, 1000/box
  • Warranty3 month
3 Month Warranty

Key Features

  • Tested for absorption of 1.8 to 2.2 mL of Culture media
  • pH neutral
  • Individually packaged to assure sterility
  • 3 month warranty

More About this Item

Absorbent pads are white cellulose discs that fit into standard petri dishes. Pre-sterilized and tested for absorption of 1.8 to 2.2 mL of culture media, these pads are ideal for culturing using broth media in petri dishes.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat