SalvisLab 31A04161 Programmable Incucenter Natural Convection Incubator, 4.2 cu ft, 115 VAC

Representative image only.
SalvisLab Mfr # 31A04161 – Item # WW-39702-10
Easy to operate and program!
  • Mechanical forced air convection provides excellent temperature uniformity
  • Natural air convection units are ideal for drying applications that require gentle air flow
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (cu ft)2.8
  • Capacity (Liters)79.3
  • Control TypeMicroprocessor
  • Max Temperature (° C)110
  • Overall Depth (in)23 5/16
  • Overall Depth (cm)59.2
  • Overall Height (in)21 5/16
  • Overall Height (cm)54.1
  • Overall Width (in)29 1/2
  • Overall Width (cm)74.9
  • Power (VAC)115
  • Chamber Depth (in)15 1/2
  • Chamber Depth (cm)39.4
  • Chamber Height (in)15 3/4
  • Chamber Height (cm)40
  • Chamber Width (in)19 1/8
  • Chamber Width (cm)48.6
  • Number Of Shelves1
  • DescriptionProgrammable Incucenter Natural Convection Incubator, 4.2 cu ft, 115 VAC
  • Warranty2 year
Two year warrantyConformite Europeenne

Key Features

  • Mechanical forced air convection provides excellent temperature uniformity
  • Natural air convection units are ideal for drying applications that require gentle air flow
  • 2 year warranty

More About this Item

Salvis Incucenter programmable mechanical forced-air incubators have the exclusive IntelliFan™ system that provides uniform temperature throughout the chamber independent of load. The maximum temperature of 110°C is ideal for decontamination procedures.

The new Salvis Incucenter programmable natural air convection incubators are ideal for
gentle incubation of organisms, such as on agar plates, and also for conditioning of heat sensitive media.

Programmable capabilities include real-time clock, timer with delayed start from 0 to 999 hours 59 minutes, RS-232 interface, variable fan speed and temperature ramp. LCD display has 5 different languages to choose from for easy programming. Built-in memory back-up retains programs even if power fails! Two separate over-temperature safety features take control of heating and shut down the incubator in the event of main controller failure. The safety glass inner door prevents heat and energy loss while samples are being monitored.

Other features include PT 100 sensor for increased accuracy and a 6 mm access port for additional temperature sensors and cables. The inner chamber is constructed of stainless steel with rounded transitions between walls to make cleaning easy. Purchase optional air filter to reduce contamination in the natural air convection units.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat