Salisbury by Honeywell SKJP8-2 Arc Flash Protective Jacket/Overpant Kits, 8 cal/cm2, double extra large

Representative image only.
Salisbury by Honeywell Mfr # SKCP82XL – Item # WW-86436-73
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Length (in)32
  • Length (cm)81.28
  • SizeXX-Large
  • ColorBlack
  • ComplianceHazard Risk Category 2
  • DescriptionArc Flash Protective Jacket/Overpant Kits, 8 cal/cm2, double extra large
  • Warranty1 year
1 Year Warranty

Key Features

  • 1 year warranty

More About this Item

Kits include jacket, overpants, helmet with face shield, safety glasses, and carrying bag.
Pro-Wear™ Jackets and Overpants features
* Made with Indura Ultra Soft® (88% cotton) for comfort and breathability
* Meet current ASTM F1506 and NFPA 70E standards
* CE 0120 certified
* Jackets feature 32" length, Nomex® wristlets, and hook & pile front closure
* Overpants feature 30" inseam, hook & pile cuff adjustment, and loose design for fitting over existing work clothes
Helmet with face shield features
* Face shield has 70% visual light transmission (VLT) — best in the industry!
* 7.5" x 20" viewing area
* Absorbs 99.9% of harmful UV radiation
* Tested to ASTM F2178 specifications

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat