Salisbury by Honeywell S23870 10 Piece Insulated Screwdriver Set
Representative image only.
Salisbury by Honeywell Mfr # S23870 – Item # WW-91100-43
Helps workers meet OSHA requirements
  • Rated for exposure up to 1000 VAC and dielectrically tested at 10,000 VAC
  • Meet or exceed ASTM 1505-07 and IEC 900 Standards for Insulated Hand Tools
  • Helps comply with OSHA 1910.333 (c)(2), and NFPA 70E 2004
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Screwdriver TypePhillips®, Slotted
  • Set IncludesNo. 1 with 3'' blade length (Phillips), No. 2 with 4'' blade length (Phillips), No.3 with 6'' blade length (Phillips), 1/8'' with 2'' blade length (slotted), 3/16'' with 6'' blade length (slotted), 3/16'' with 10'' blade length (slotted), 1/4'' with 4'' blade length (slotted), 5/16'' with 6” blade length (slotted), 3/8'' with 8'' blade length (slotted), 3/8'' with 12'' blade length (slotted), Cloth and vinyl roll with snap closure
  • Number of Pieces per Set10
  • Description10 Piece Insulated Screwdriver Set
  • Warranty1 Year
1 Year Warranty
Key Features
  • Rated for exposure up to 1000 VAC and dielectrically tested at 10,000 VAC
  • Meet or exceed ASTM 1505-07 and IEC 900 Standards for Insulated Hand Tools
  • Helps comply with OSHA 1910.333 (c)(2), and NFPA 70E 2004
  • 1 year warranty
More About this Item
These quality insulated tools are made with impact-resistant and flame-resistant material. Two-color insulation makes inspection easier which adds to the overall safety. If the yellow under layer is showing, the tool may no longer be insluated properly and should be considered for replacement and shall be removed from service.
Custom kits and individual tools available. Call our Application Specialists for more information.
10 Piece Insulated Screwdriver Set
No. 1 with 3'' blade length (Phillips)
No. 2 with 4'' blade length (Phillips)
No.3 with 6'' blade length (Phillips)
1⁄8'' with 2'' blade length (slotted)
3⁄16'' with 6'' blade length (slotted)
3⁄16'' with 10'' blade length (slotted)
1⁄4'' with 4'' blade length (slotted)
5⁄16'' with 6” blade length (slotted)
3⁄8'' with 8'' blade length (slotted)
3⁄8'' with 12'' blade length (slotted)
Cloth and vinyl roll with snap closure