Safetec 15205 Hazardous Lab Chemical Spill Clean-Up Kit
Representative image only.
Safetec Mfr # 15205 – Item # WW-86105-04
Neutralize, stabilize, and solidify dangerous acids, bases, aqueous solutions, and organics
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHazardous Lab Chemical Spill Clean-Up Kit
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Kit includes, wall-mount dispenser, RED Z, one pair of nitril acid-resistant gloves, pick-up scoop with scraper, waste bags (6), hazardous and non-hazardous labels, GREEN Z, PETRO LOCK, ACID LOCK, ALKY, hazardous material accident reporting kit.