Rotromoc PS14 Replacement Sample Cup for Fast Benchtop System, 14 mm
Representative image only.
Rotromoc Mfr # PS14 – Item # WW-37910-67
Prevents soiling or cross contamination
  • Provides convenient means of collecting and storing samples
SPECS & DESCRIPTION
Specifications & Description
  • ColorTransparent
  • Overall Height (in)1 3/4
  • Overall Height (cm)4.4
  • DescriptionReplacement Sample Cup for Fast Benchtop System, 14 mm
Key Features
  • Provides convenient means of collecting and storing samples
More About this Item
The disposable sample containers ensure the optimum sample volume is introduced into the Rotronic WP-14-S sample holders. Prevents sample holders from coming into direct contact with the product being tested, thereby preventing soiling or cross contamination. The sample containers also provide a convenient means of collecting and storing samples.