Replacement glass beads, 2 mm dia., 10 oz per bag
Representative image only.
Market Forge Item # WW-10779-50
₹10,500.00INR / Bag of 10
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionReplacement glass beads, 2 mm dia., 10 oz per bag
More About this Item
Keep your sterilizer working at its best with these parts and accessories.
Replacement glass beads for Dry Glass Bead Sterilizers. Beads are 2mm in diameter. One 10 oz bag refills the 3" diameter sterilizer twice, and the 5" diameter sterilizer once.