Ralston Instruments QTHA-2FSA-RG-QD Ralston , SS Adapter no Check; 1/4"BSPP(F) to Quick-Test(F)
Representative image only.
Ralston Instruments Mfr # QTHA-2FSA-RG-QD – Item # WW-16107-11
Leak free connections for pressure calibrations
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRalston , SS Adapter no Check; 1/4\"BSPP(F) to Quick-Test(F)
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
High quality fitting kits and connection hoses are designed to give reliable and leak free connections for pressure calibrations. Since these fittings do not require wrenches or thread sealant, these fitting kits and quick connect hoses reduce calibration time and down time due to calibration of pressure gauges and transmitters.
The QTHA series of adapters listed all have a Quick-test (female) end to connect to the benchtop manifolds. The other end has some of the more popular connection types for adapting to your application. Stainless steel construction with Buna N sea