Ralston Instruments QSQS-HOS-20FT Quick-test Stainless Steel Hose 6 ft
Representative image only.
Ralston Instruments Mfr # QSQS-HOS-20FT – Item # WW-16109-20
Leak free connections for pressure calibrations
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionQuick-test Stainless Steel Hose 6 ft
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
High quality fitting kits and connection hoses are designed to give reliable and leak free connections for pressure calibrations. Since these fittings do not require wrenches or thread sealant, these fitting kits and quick connect hoses reduce calibration time and down time due to calibration of pressure gauges and transmitters.
The microbore hose provides a quick, low-volume, high-pressure way of connecting any pressure source to an instrument under test. The very small microbore volume reduces pumping effort and the amount of compressed gas needed during calibration. Both ends terminate with Quick-test stainless steel connectors that need no tools to tighten. A corrosion-resistant strain relief on each end reduces wear and dramatically impacts hose life. The hose is pinpricked—ok for use in natural gas applications.