Ralston Instruments 742129 5 piece brass adapter kit
Representative image only.
Ralston Instruments Mfr # QTHA-KIT0 – Item # WW-68873-66
Leak free connections for pressure calibrations
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Description5 piece brass adapter kit
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
High quality fitting kits and connection hoses are designed to give reliable and leak free connections for pressure calibrations. Since these fittings do not require wrenches or thread sealant, these fitting kits and quick connect hoses reduce calibration time and down time due to calibration of pressure gauges and transmitters.