Rainwise PORTLOG Portable Weather Station, solar-powered
Representative image only.
Rainwise Mfr # PORTLOG – Item # WW-99755-06
Designed to measure and record in remote areas
  • Log up to one year of weather measurements
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Wind Speed (mph)150
  • Min Wind Speed (mph)0
  • Max Temperature (Outside) (° F)150
  • Min Temperature (Outside) (° F)-66
  • Max Temperature (Outside) (° C)65
  • Min Temperature (Outside) (° C)-54
  • Max Humidity RH (Outside) (%)100
  • Min Humidity RH (Outside) (%)0
  • Wind Speed Accuracy±2% full-scale
  • DescriptionPortable Weather Station, solar-powered
  • Warranty5 years
5 Year Warranty
Key Features
  • Log up to one year of weather measurements
  • 5 year warranty
More About this Item
This solar powered, self-contained weather station measures and records barometric pressure, temperature, humidity, dew point, wind direction, wind speed, rain fall and solar radiation. Ideal for remote field measurements. Program the logging sample rate from once a minute to once an hour. At one sample per hour, the datalogger will store over one year of measurements. Download data to a laptop for evaluation with included software and RS-232 interface. Weather station fits into rugged transport case.