Qsonica 4421 COUPLER; USE WITH 04712-65
Representative image only.
Qsonica Mfr # 4421 – Item # WW-04712-64
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Replacement Probe selection is determined by sample volume, sample type, and the desired intensity. As tip diameter increases, the intensity/amplitude decreases. Tips erode over time and replacement tips are available for order.