Qsonica 4121 Booster for Ultrasonic Processors, 2:1 gain
Representative image only.
Qsonica Mfr # 4121 – Item # WW-04712-56
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Tip Diameter (cm)0
  • DescriptionBooster for Ultrasonic Processors, 2:1 gain
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Customize your ultrasonic processor with these quality accessories and replacements.
Attach between the converter and probe. Booster doubles the intensity of standard ¨ú¡È and 1¡È probes. Reduces processing times by up to 50%, and is ideal for processing larger sample volumes.