QRP VHC12 XLARGE Vinyl Cleanroom Gloves, extra large, 100/bag
Representative image only.
QRP Mfr # VHC12 XLARGE – Item # WW-86580-23
Perfect for pharmaceuticals, electronics, and cleanroom food processing
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)6
  • MaterialVinyl
  • SizeX-Large
  • Length (in)12
  • Length (cm)30.5
  • ColorWhite
  • GripMicro-textured
  • ComplianceISO Class 5; USDA; IEST-RP-CC-005.2
  • SterileNo
  • DescriptionVinyl Cleanroom Gloves, extra large, 100/bag
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
Gloves feature a micro-textured surface for enhanced gripping. Latex-free, 100% PVC co-polymer is naturally antistatic with a resistivity of 1010 ohms. Beaded cuff for additional strength. Ambidextrous. Packed 100 gloves per polybag.
Compliance: ISO Class 5, USDA, and IEST-RP-CC-005.2.