QLA UltraCenter™ Precision Vessel Compatible with Agilent 708/709-DS Baths, Clear Glass with Plastic Rim, 1000 mL; Serialized
Representative image only.
Mfr # ASTM900-70XDS – Item # WW-41301-01
Get better dissolution results with UltraCenter precision glass vessels
  • Glass offers good thermal conductivity and high resistance to scratching
  • Serialized—include a Certificate of Conformance (CoC)
  • For Agilent 708/709-DS
  • Clear with plastic rim
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)1000
  • Capacity (oz)33.8
  • MaterialGlass
  • Bottle ColorClear
  • Special FeaturesPlastic rim
  • Outside Diameter (OD) (in)4 7/32
  • Outside Diameter (OD) (cm)10.725
  • DescriptionUltraCenter™ Precision Vessel Compatible with Agilent 708/709-DS Baths, Clear Glass with Plastic Rim, 1000 mL; Serialized
Key Features
  • Glass offers good thermal conductivity and high resistance to scratching
  • Serialized—include a Certificate of Conformance (CoC)
  • For Agilent 708/709-DS
  • Clear with plastic rim
More About this Item
UltraCenter Precision glass vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius, resulting in more reproducible dissolution test results. They are compatible with most dissolution baths. Each vessel features a unique serial number.
Glass vessels with plastic rims reduce chipping and cracking, minimizing glass breakage. Vessels are laser marked on plastic rims with unique serial numbers.
Not available for sale in the United States.