QLA Basket Sinker, 316 SS; Japanese Pharmacopeia
Representative image only.
Mfr # CUSBSK-JP – Item # WW-41308-55
Capsule sinkers improve reproducibility
  • Keep floating tablets from sticking to sides of vessels
  • 0.47" (11.9 mm) ID
  • Japanese Pharmacopeia
PRODUCT OPTIONS
₹35,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Material316 stainless steel
  • Diameter (in)0.47
  • Diameter (mm)12
  • DescriptionBasket Sinker, 316 SS; Japanese Pharmacopeia
Key Features
  • Keep floating tablets from sticking to sides of vessels
  • 0.47" (11.9 mm) ID
  • Japanese Pharmacopeia
More About this Item
Sinkers help to ensure tablets dissolve at more reproducible rates and keep them from sticking to the sides of vessels. Select from spring style, basket style, three-prong style, and others.
Not available for sale in the United States.