Pyrex 9985-100 Brand 9985 watch glass; 100 mm, case of 144
Representative image only.
Pyrex Mfr # 9985-100 – Item # WW-34507-24
SPECS & DESCRIPTION
Specifications & Description
  • Fits Bottle / Beaker Size (mL)600
  • MaterialGlass
  • AutoclavableYes
  • Outside Diameter (OD) (cm)10
  • DescriptionBrand 9985 watch glass; 100 mm, case of 144
More About this Item
These watch glasses provide high chemical durability, preventing contamination of solutions. They are modeled to the same radius of curvature for convenient stacking. The heavy wall an uniform fire-polished edge provide mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected to occur.