Pyrex 3025-250 Cylinder, Brand 3025, graduated, 250 mL
Representative image only.
Pyrex Mfr # 3025-250 – Item # 3454605
Double metric scale, economy
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)250
  • Graduation Interval (mL)2
More About this Item
These double metric scale cylinders are calibrated "to contain." These economical cylinders are ideal for many general laboratory procedures.
Graduated ware must carry markings listing its capacity and the temperature at which it should be used, and whether the piece was calibrated T.C., "to contain," or T.D., "to deliver," the stated volume. T.C. calibration indicates the volume held in the co