Pyrex 2982-50 Brand 2982 cylinder; 50 mL, case of 12
Representative image only.
Pyrex Mfr # 2982-50 – Item # 3454523
Feature single metric scale, PYREX stopper
PRODUCT OPTIONS
Call For Price
₹17,600.00 - ₹25,900.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Capacity (mL)50
  • Graduation Interval (mL)1
  • Tolerance (mL)±0.50
  • Cap TypeStopper
  • Cap MaterialGlass
More About this Item
These graduated cylinders are calibrated "to contain" and are useful as mixing cylinders. Each cylinder features the PYREX stopper. Graduations are in durable white enamel.
When taper sizes are given, the first number is the stopper diameter (mm) across the top; the second number is the stopper length (mm). If only one number is given, it indicates the stopper diameter (mm) across the top.