Cole-Parmer Essentials Spatula with .5" Spoon End x 1.5" Flat Round End, PTFE Coated Stainless Steel
Representative image only.
Cole-Parmer Essentials Item # WW-06369-10
Handle samples easier
  • Prevent build-up with PTFE coating
  • Won't scratch or mar delicated surfaces
  • 1 year warranty
PRODUCT OPTIONS
₹5,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)6 1/4
  • Overall Length (cm)15.88
  • MaterialPTFE-coated Stainless Steel
  • SterileYes
  • DescriptionPTFE-coated lab spatula, with 1/2\"L spooned end and 1 1/2\"L flat rounded end
  • Warranty1 year
1 Year Warranty
Key Features
  • Prevent build-up with PTFE coating
  • Won't scratch or mar delicated surfaces
  • 1 year warranty
More About this Item
Double-ended spatulas are completely covered with extra heavy-duty coating of PTFE—prevents buildup on surfaces or loss of critical quantities due to sticking. Ideal for handling chemicals.