Pro Spore PA7953105 Ampule Self Contained Steam Bio Indicator 105 Population, 50 Amp/pack
Representative image only.
Pro Spore PA7953105 Ampule Self Contained Steam Bio Indicator 105 Population, 50 Amp/pack
Pro Spore | Mfr: PA7953105| Item: 9470230
Call For Price
Call For Price
Product Description
More About this Item
ProSpore Ampoule is a self-contained Biological Indicator available for use in steam sterilization. It is ideal for use in validating the sterilization of volumes of liquid that take a longer time to reach sterilizing conditions. The Ampoule is filled with modified Soyabean, Casein digest broth containing bromocresol purple (acid) indicator. Complies with EN 866-7 and AAMI ST 19.
Specifications
  • DescriptionAmpule Self Contained Steam Bio Indicator 105 Population, 50 Amp/pack