Precision Digital PD684-0K1 Process meter, general purpose

Representative image only.
Precision Digital Mfr # PD684-0K1 – Item # WW-61600-02
Intrinsically safe model for use in just about any hazardous area.
  • 5-digit backlit display with bargraph
  • Open collector alarm or pulse output
  • HART protocol transparent
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • DIN Size1/8\"
  • Control Input mA4-20
  • Display Type0.6\" backlit LCD
  • Display Digits5
  • Max Operating Temperature (° F)149
  • Min Operating Temperature (° F)-4
  • EnclosureHigh-impact plastic, uL 94V-0
  • Overall Width (in)4 43/64
  • Overall Height (in)2 29/64
  • Overall Depth (in)3 51/64
  • Overall Width (mm)118.9
  • Overall Height (mm)62.2
  • Overall Depth (mm)96.3
  • DescriptionProcess meter, general purpose
  • Warranty3 Years
3 Year WarrantyConformite EuropeenneCanadian Standards AssociationFM ApprovedIntrinsically Safe

Key Features

  • 5-digit backlit display with bargraph
  • Open collector alarm or pulse output
  • HART protocol transparent
  • 3 year warranty

More About this Item

The loop-powered rate/totalizers can display rate up to five digits and total up to eight digits using the total overflow feature. The display can be programmed to automatically or manually toggle between rate and total. Standard features include a full 5-digit backlit display with bargraph, on-screen custom engineering units, and max/min display capabilities.

Square root and ptrogrammable exponent functions are standard features on these meters. These indicators can display flow rate and total from a DP transmitter or open channel flow in weirs and flumes. Programming is easy through the fron panel buttons, with no pots or jumpers to deal with, and can be scaled without a signal source.

The Loop Leader Series meters go just about anywhere. The PD689, with FM, CSA and ATEX certifications, can be installed in just about any hazardous area.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat