Porta-Lab TAB-2012 Touch Activated Laboratory Micro Burner
Representative image only.
Porta-Lab Mfr # TAB-2012 – Item # WW-36310-56
Touch-Activated Fuel Type Butane
  • Hands free operation
  • Safety On-Off Lock
  • Auto On-Off Ignition
  • Adjustable flame control
  • Unit size: 5"X2"X2.25"
  • FueL Capacity: 35gms
  • Max.Temp: 2500°F
  • 1 year warranty
₹10,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Gas TypeButane
  • DescriptionTouch Activated Laboratory Micro Burner
  • Warranty1 year
1 Year Warranty
Key Features
  • Hands free operation
  • Safety On-Off Lock
  • Auto On-Off Ignition
  • Adjustable flame control
  • Unit size: 5"X2"X2.25"
  • FueL Capacity: 35gms
  • Max.Temp: 2500°F