This item is now discontinued. We recommend the following as suggested replacements: WW-03030-51
Porta-Lab PRO-2121 Laboratory Handheld Torch/Portable Burner; Butane
Representative image only.
Porta-Lab Mfr # PRO-2121 – Item # WW-36310-41
Use as both a handheld torch and portable burner
  • Butane fuel is self-contained and can be refilled with standard butane canister fuel
  • Single-knob gas control with trigger-activated electronic auto ignition
  • One fuel canister provides over 2500°F (1371°C) and up to 1.5 hours of continuous burn time
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Gas TypeButane
  • Width (in)3 1/2
  • Length (in)5 1/2
  • Height (in)8
  • DescriptionLaboratory Handheld Torch/Portable Burner; Butane
  • Warranty1 Year
1 Year Warranty
Key Features
  • Butane fuel is self-contained and can be refilled with standard butane canister fuel
  • Single-knob gas control with trigger-activated electronic auto ignition
  • One fuel canister provides over 2500°F (1371°C) and up to 1.5 hours of continuous burn time
  • 1 year warranty
More About this Item
This unit can be used as both a torch and a burner. The torch can be inserted in the provided black base or turned on its side with nozzle pointing directly upward resting on the included metal ring
stand to simulate a benchtop burner.
This affordable torch burner contains a completely controlled flame size that can be manually controlled for off/on or set for a continuous flame. Because the unit will not flare in any position,
this torch can be used as a portable bunsen burner when used with included metal stand. There is an ON/OFF switch for safety. Compact size (5 1⁄2''(14 cm) L x 3 1⁄2''(9 cm) W x 8''(21 cm) H) make this an excellent burner for field use.