This item is now discontinued. We recommend the following as suggested replacements: WW-78930-17
PolyScience Bath Fluid, Ethylene Glycol Solution, -25 to 100 C, 1/2 Gal; 5/CS
Representative image only.
PolyScience Mfr # 004-300060 – Item # WW-01294-31
Get more out of your bath!
  • Maintains bath temperature as high as 280°C
  • 0ver temperature cutoff provides added operator safety
  • 2-1/2 Years Parts and Labor Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Temperature (° C)100
  • Min Temperature (° C)-25
  • DescriptionBath Fluid, Ethylene Glycol Solution, -25 to 100 C, case of five 1/2-gal
  • Warranty2-1/2 Years Parts and Labor
2 and a Half Year Warranty
Key Features
  • Maintains bath temperature as high as 280°C
  • 0ver temperature cutoff provides added operator safety
  • 2-1/2 Years Parts and Labor Warranty
More About this Item
Use bath fluids to maintain desired bath temperatures, outside of the range for water. For operator safety, set the overtemperature cutoff of your bath/circulator below the flash point of the bath fluid.
Note: We do not recommend using automotive anti-freeze with circulating baths — additives can be harmful to the circulating pump. Silicone bath fluids are not compatible with silicone tubing.