Platinum Cover for 40 mL Standard Form Crucibles, 1/ea

Representative image only.
Mfr # GCS40LID – Item # WW-17831-65
Prohibit contamination at high temperatures up to 2192°F (1200°C)
  • Constructed of durable platinum

PRODUCT OPTIONS

Call For Price

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPlatinum
  • Capacity (mL)40
  • Cap MaterialPlatinum
  • AutoclavableYes
  • DescriptionPlatinum Cover for 40 mL Standard Form Crucibles, 1/ea

Key Features

  • Constructed of durable platinum

More About this Item

Platinum offers oxidation resistance in air and oxygen; and corrosion resistance in acids and melted salts. Lids, crucibles, and tongs sold separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat