Pipette Tracking System

Representative image only.
Item # WW-26000-00
No more pipetting errors
  • Automatically detects, guides, and records your pipetting dispense steps
  • Optical sensors detect actual pipetting locations and flag them with lights and sound
  • Versatile — compatible with standard 96-/384-well microplates and tube strips
  • Ensure quality control with detailed log file
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • SterileNo
  • Overall Length (in)10
  • Memory8 GB NAD Flash, 64 GB MicroSD (not included)
  • Power (VAC)100 to 240
  • BatteryBuilt-in 3.7 V battery
  • Compatible withPlates & Tubes: 384-well (height <11 mm), 96-well (height <23.5 mm), 8-tube and 12-tube PCR strips, ELISA and storage microplates
    Tips: 0.1 to 1000 µL
  • DescriptionPipette Tracking System
  • Warranty1 year
1 Year Warranty

Key Features

  • Automatically detects, guides, and records your pipetting dispense steps
  • Optical sensors detect actual pipetting locations and flag them with lights and sound
  • Versatile — compatible with standard 96-/384-well microplates and tube strips
  • Ensure quality control with detailed log file
  • 1 year warranty

More About this Item

This tablet-driven system eliminates the need for you to input any pipetting protocol or to press an advance button after every dispense. Unique system uses scanning infrared LED lights to detect pipetting steps and displays them on the tablet''s preloaded application as a dot, which changes color with each new dispense. The dots appear in both the sample detection and the control windows, and show the number of times each well received a dispensed amount. Two sensor scanning layers accommodate both 96- and 384-well microplates, as well as PCR tube strips (optional adapter required).


Set up is easy! Choose the pipettor type (single, 8, or 12 channel, select the detection sensitivity and plate type, initiate the session, and begin pipetting. The last location blinks on the screen, so you don''t lose your place.


Each pipetting step is recorded in a log file with time and location, providing a detailed record for analysis and quality control. Imbedded NAD Flash (8 GB) memory stores data for later download to PC. Or use an external storage device — MicroSD port supports up to 64 GB.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat