PCRmax Eco 48 Real-Time qPCR System; 100 to 240 VAC

Representative image only.
PCRmax Item # WW-93947-00
Patented block and optical technologies deliver extreme speed, sensitivity, and data quality
  • HRM functionality as standard with the ability to discriminate class IV SNP >99% of the time
  • Eco 48 allows for simple multiplex analysis with up to four dyes at once
  • World's most thermally accurate block gives consistent data every time
  • High uniformity provides high quality data. Fast cycling enables several experiments per day, all at an economical price
  • Fastest block-based real-time PCR system: 40 cycles in 40 minutes as standard and 40 cycles in as little as 15 minutes after optimization of temperatures and hold times
  • Open platform that can utilize any chemistry, dye or PCR reagent
  • Easy to use software, making analysis simple and fast
  • Provides a small 34.5cm x 31cm (13.6” × 12.2”) footprint to save bench space
  • ECO 48 thermal technology
  • Research Use Only
  • 3 year warranty

Call For Price

₹377,000.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Max Temperature (° C)100
  • Min Temperature (° C)30
  • Temperature Accuracy±0.25⁰C
  • Ramp RateAverage: 5.5⁰C per second
  • Block Uniformity±0.1⁰C
  • Block DescriptionProprietary hollow silver/gold block, Peltier-based system with conductive fluid, 48 wells
  • MemoryUnit saves the run data of last experiment, or unlimited via USB
  • Length (in)13 5/8
  • Height (in)12 5/8
  • Width (in)12 13/64
  • Length (cm)34.5
  • Height (cm)32
  • Width (cm)31
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • DescriptionReal-Time qPCR System; 100 to 240 V, 50/60 Hz
  • Warranty3 Years
3 Year Warranty

Key Features

  • HRM functionality as standard with the ability to discriminate class IV SNP >99% of the time
  • Eco 48 allows for simple multiplex analysis with up to four dyes at once
  • World's most thermally accurate block gives consistent data every time
  • High uniformity provides high quality data. Fast cycling enables several experiments per day, all at an economical price
  • Fastest block-based real-time PCR system: 40 cycles in 40 minutes as standard and 40 cycles in as little as 15 minutes after optimization of temperatures and hold times
  • Open platform that can utilize any chemistry, dye or PCR reagent
  • Easy to use software, making analysis simple and fast
  • Provides a small 34.5cm x 31cm (13.6” × 12.2”) footprint to save bench space
  • ECO 48 thermal technology
  • Research Use Only
  • 3 year warranty

More About this Item

The PCRmax® Eco48 real time PCR system is a high specification, economically priced real time thermal cycler that accommodates a unique 48-well polypropylene PCR plate utilizing the same geometry as standard 384-well plates, but only 1/8 of the size. This enables users to dramatically reduce the qPCR reagent volumes compared to traditional 96-well instruments, whilst still producing a strong fluorescence signal. Minimizing the plate size also significantly improves thermal uniformity. A minimum volume of 5 μl is validated, cutting reagent costs in half and making more efficient use of expensive and hard to acquire template DNA samples. MIQE compliant open license software gives confidence when it comes to publishing your data. Eco48 allows for simple multiplex analysis with up to four dyes at once. Fast cycling enables several experiments per day, all at an economical price. Eco48 is an open platform that can utilize any chemistry, dye or PCR reagent. The easy to use software, makes analysis simple and fast.

The Eco48 system comes with two distinct softwares, the first, Eco Control, drives the unit's set up and experimental runs while Eco Study is the analysis package. The Eco48 software is open license, you can install them on as many PCs as you like, and MIQE compliant. With the Eco 48 system and software, data collection is monitored in real time, allowing researchers to access run viability immediately. The user-friendly data analysis interface also allows researchers to easily view the amplification plot, melt curve analysis, and the analyzed results, including Cq values, PCR efficiency, R2 and Y-intercept.

Related Article: Using PCR and qPCR in Water Treatment

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat