Palintest CS 100 Chlorosense Chlorine Meter

Representative image only.
Palintest Mfr # CS 100 – Item # WW-99359-10
Chlorine measurement without powders, liquids, or tablets – no chemical waste
  • Waterproof, rugged, and portable
  • Reads up to 100 ppm chlorine
  • Unique sensor technology
  • USB port for easy connection
  • Up to 500 results stored
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min ISE0.02 to 100 ppm
  • ISE Accuracy0.01 to 10 ppm
  • Min Temperature (° C)2
  • Max Temperature (° C)40
  • Min Temperature (° F)35
  • Max Temperature (° F)104
  • InterfaceUSB
  • WaterproofYes
  • Battery4 x AA batteries (included)
  • Width (cm)16.98625
  • Height (cm)11.58875
  • Width (in)6 11/16
  • Length (in)5
  • Height (in)4 9/16
  • DescriptionChlorosense Chlorine Meter
  • Warranty1 year
1 Year WarrantyConformite Europeenne

Key Features

  • Waterproof, rugged, and portable
  • Reads up to 100 ppm chlorine
  • Unique sensor technology
  • USB port for easy connection
  • Up to 500 results stored
  • 1 year warranty

More About this Item

ChloroSense uses a unique sensor to measure free and total chlorine simultaneously in potable water samples. The electrochemical sensors are individually packed and sealed. There are no chemical reagents to handle and measure out. Each pack of sensors comes with its own unique calibration chip matched to the sensors at time of manufacture.
Conducting a test is easy. Open the instrument and fill the sample container with water; open and insert the sensor; close the instrument and the test automatically begins. Free and total chlorine results are displayed along with temperature. The chlorine readings are automatically adjusted for temperature. Results (up to 500) are stored for later recall.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat