This item is now discontinued. We recommend the following as suggested replacements: WW-63528-72
Pipettor Rack, clear, holds 6 pipettors
Representative image only.
Item # WW-07952-75
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • DescriptionPipettor Rack, clear, holds 6 pipettors
More About this Item
Maximize bench space with these sturdy pipettor stands. Model 07952-70 holds up to six pipettors along with a tray pf prepackaged pipette tips. Benchtop PLEXIGLAS rack 07952-75 holds up to six pipettors diagonally (not for use with BENCHMATE pipettors). Use stands to store pipettors when not in use.