Onguard Chemical Splash Suit, Small
Representative image only.
Onguard Mfr # 71022-S – Item # WW-86234-00
Best value in chemical splash protection
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • SizeSmall
  • MaterialPolyvinyl Chloride (PVC);Nylon;Polyester
  • ColorGreen
  • ComplianceFDS 5903.1
  • DescriptionChemical Splash Suit, Small
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Splash suits consist of coveralls with an attached hood made from 0.35-mm heavy-duty PVC on nylon/polyester. Features include reinforced crotch, elbows, and knees; heavy-duty elastic suspender with clip; adjustable waist snaps; heavy-duty zipper; cord locks on hood strings; and ankle and cuff take-up snaps for a comfortable fit. Wallet size degradation chart is included. For use in Level C environments.
Compliance: Meets Federal Apparel Flammability Act, FDS 5903.1