Ohio Semitronics ACT-100E Single Phase AC Current Transducer, 0 to 100 A
Representative image only.
Ohio Semitronics Mfr # ACT-100E – Item # WW-68471-74
Designed for use in applications where UL listed measurement devices are required
  • Accurate, low cost, reliable current measurement
  • Designed to withstand motor start-up transients
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Input (AAC)100
  • Min Input (AAC)0
  • Max Output (ADC)0.02
  • Min Output (ADC)0.004
  • Max Supply Power (VAC)115
  • Basic Accuracy (%)±0.25% full-scale @ 60 Hz (internal sensor), ±0.5% full-scale @ 60 Hz (external sensor)
  • DescriptionSingle Phase AC Current Transducer, 0 to 100 A
  • Warranty5 Years
5 Year Warranty
Key Features
  • Accurate, low cost, reliable current measurement
  • Designed to withstand motor start-up transients
  • 5 year warranty