Smart sensor heads retain calibration and sensor information to optimize time and efficiency
Extremely versatile—the meter automatically recognizes which sensors you plug in so you can easily switch parameters whenever you want
Select a meter with the number of channels you need (1, 2, or 3) and the sensors for the parameters you are measuring (pH, conductivity, or DO)
Eliminate downtime with smart sensors that feature built-in calibration and measurement information
Rugged, waterproof and dustproof housing with an IP67 rating for use in any environment
Nonslip ergonomic design is comfortable to use and shock-proof and scratch-resistant
Easily view measurements with large color graphic TFT-LCD, even in dim lighting or bright outdoor sunlight
Download and analyze up to 10,0000 data points easily with micro USB
Accurately measure pH, ORP, conductivity, TDS, resistivity, salinity, dissolved oxygen, and temperature with automatic sensor recognition function, sensors sold separately
Connectionsmicro USB, phono jack, and push-pull connection
Min pH (pH)-2
Max pH (pH)20
pH Resolution0.001
pH Accuracy±0.005
Buffer RecognitionUSA, DIN, NIST, NIST (10), and custom
CalibrationpH: Up to 6 points; Conductivity: Up to 4 automatic, up to 5 manual
Min mV (mV)-1000
Max mV (mV)1000
mV Resolution0.1
mV Accuracy±0.1
Min Conductivity0 S
Max Conductivity200 S
Conductivity ResolutionAuto ranging, up to 4 significant digits
Conductivity Accuracy±0.5% full scale of each range, ±1.5% full scale at 200 mS
Min TDS0.01 mg/L
Max TDS200,000 mg/L
TDS Resolution0.01 minimum, 4 significant digits
TDS Accuracy±0.5% of reading or ±0.1 mg/L, whichever is greater
Min Salinity0 ppt
Max Salinity80 ppt
Salinity Resolution0.01 ppt
Salinity Accuracy±0.5% of reading or ±0.01 ppt, whichever is greater
Min Resistivity0.1 Ω•cm
Max Resistivity200 MΩ•cm
Resistivity ResolutionAuto ranging, up to 4 significant digits
Resistivity Accuracy±0.5% full scale of each range, ±1.5% full scale > 20.0MΩ
Min Dissolved Oxygen0 mg/L, 0%
Max Dissolved Oxygen20 mg/L, 200%
Dissolved Oxygen Resolution0.01 mg/L, 0.1%
Dissolved Oxygen Accuracy±0.2 mg/L,±0.2%
Min Temperature (° C)-30
Max Temperature (° C)130
Min Temperature (° F)-22
Max Temperature (° F)266
Temperature Resolution0.1°F/°C
Temperature Accuracy±0.9°F (±0.5°C)
Temperature compensationAutomatic
Temperature Coefficient0.00 to 10.00%/°C
InterfaceUSB and Wi-Fi
Outputmicro USB for PC connection, phono jack for printer connection
Data Logging (points)10000
WaterproofIP67
Battery4 AA (included)
Width (cm)3.45
Height (cm)22
Width (in)1 2/5
Height (in)8 21/32
Number of Positions3
NIST Calibration IncludedNo
Probe IncludedNo
Accessories Includedfour AA batteries, and wireless SD card
DescriptionWaterproof 3-Channel Smart Handheld Meter
Warranty3 year
Includes
meter, four AA batteries, and wireless SD card.
Key Features
Extremely versatile—the meter automatically recognizes which sensors you plug in so you can easily switch parameters whenever you want
Select a meter with the number of channels you need (1, 2, or 3) and the sensors for the parameters you are measuring (pH, conductivity, or DO)
Eliminate downtime with smart sensors that feature built-in calibration and measurement information
Rugged, waterproof and dustproof housing with an IP67 rating for use in any environment
Nonslip ergonomic design is comfortable to use and shock-proof and scratch-resistant
Easily view measurements with large color graphic TFT-LCD, even in dim lighting or bright outdoor sunlight
Download and analyze up to 10,0000 data points easily with micro USB
Accurately measure pH, ORP, conductivity, TDS, resistivity, salinity, dissolved oxygen, and temperature with automatic sensor recognition function, sensors sold separately
3 year warranty
More About this Item
Get easy-to-read and accurate readings with auto-hold, auto-stable, and real-time measurements with stability indicators. The Environmental Express® Oakton® 300 series multiparameter meters utilize smart sensor technology with sensor heads that feature a built-in digital circuit for automatic sensor recognition. Sensor information including sensor ID, calibration data, and measurement setup is stored in sensor head and viewable on the meter display when sensor is connected.
Sensor heads are easy to connect and disconnect with waterproof push-lock connector. Sensor cartridges (pH and EC) are replaceable for a cost-efficient and environmentally friendly system. Blue LED light on the sensor head functions as a reading stability indicator, flashing until the reading is stable.
Meter stores up to 10,000 data sets with date-and-time stamp for accurate and easy analysis. Quickly transfer data from meter to computer seamlessly via USB cable or wirelessly via Wi-Fi.
Easily take measurements and conveniently store meter and electrodes with built-in foldable stand on the back of the meter. Three grooves on the meter serve as electrode holders, to hold up to three electrodes simultaneously.
Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.