Oakton Waterproof pH Electrode with ATC, Double-Junction, Epoxy-Body, Gel-Filled
Representative image only.
Oakton Item # WW-35808-72
Oakton offers a variety of pH electrodes for your general purpose and speciality laboratory applications
  • Choose single-junction electrodes to measure pH in clean aqueous samples
  • Choose double-junction electrodes for use with samples that may be contaminating with particulates, heavy metals or organic compounds
  • 1 year warranty
PRODUCT OPTIONS
₹16,100.00INR / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionGel-filled
  • Temperature CompensationYes
  • ATC Connector6-pin waterproof
  • ConnectorBNC
  • Descriptiondouble-junction, epoxy-body, gel-filled waterproof pH electrode with ATC
  • Warranty1 year
1 Year Warranty
Key Features
  • Choose single-junction electrodes to measure pH in clean aqueous samples
  • Choose double-junction electrodes for use with samples that may be contaminating with particulates, heavy metals or organic compounds
  • 1 year warranty
More About this Item
Low-maintainance epoxy-bodied electrodes provide the durability for field work. Refillable electrodes provide greater accuracy and faster response for more critical analytical work.
Compatible with Oaktoh pH 300/310, pH/CON 300, pH/CON 510, 150, & 450 series meters.