Oakton Traceable® One-Shot™ Buffer Solution, Yellow, pH 7.000; 6 x 100 mL Vials

Representative image only.
Oakton Item # WW-98767-78
Traceable® pH Buffers are Certified Reference Materials, making them one of the most accurate available
  • All Oakton® Traceable® pH buffer solutions are NIST-traceable reference materials
  • Supplied with calibration report from an ISO 17034 and ISO 17025 accredited laboratory
  • Individually packed in foil pouch to prevent UV exposure that may degrade accuracy
  • Buffer solution lets you calibrate pH instruments to ensure accuracy
  • If unopened, shelf life is 2 years
  • Use to calibrate pH instruments to 7.000
  • Color-coded buffer for easy identification

PRODUCT OPTIONS

₹4,200.00INR / Pkg of 6

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Value7
  • Buffer Kit Values7.000
  • Volume (mL)100
  • ColorYellow
  • DescriptionTraceable® One-Shot™ Buffer Solution, Yellow, pH 7.000; 6 x 100 mL Vials
NIST Traceable Calibration

Key Features

  • All Oakton® Traceable® pH buffer solutions are NIST-traceable reference materials
  • Supplied with calibration report from an ISO 17034 and ISO 17025 accredited laboratory
  • Individually packed in foil pouch to prevent UV exposure that may degrade accuracy
  • Buffer solution lets you calibrate pH instruments to ensure accuracy
  • If unopened, shelf life is 2 years
  • Use to calibrate pH instruments to 7.000
  • Color-coded buffer for easy identification

More About this Item

• Convenient, single-use vials eliminate concerns of cross-contamination; each pack contains six polyethylene vials with 100-mL of pH buffer per vial
• If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days

Oakton® Traceable® One-Shot™ pH Buffer Solutions are supplied with NIST-traceable calibration reports stating accuracy at 25°C to ±0.010 pH, in addition to conversion values for pH/temperature—for the most accurate calibrations!

METHODS SUPPORTED
• Certified reference material meets test requirements for Federal, State, and local agencies, CAP, ASTM, NCCLS, CLSI, ACS, CLIA, AOAC, EPA, APHA, AWWA, WEF, USGS, USP, and ISO. Traceable® Conductivity Standards complies with and is essential for use in these official methods: AOAC 973.40, EPA 120.1, Standard Method 2510 (APHA, AWWA, WEF), ISO 7888, DIN 38404, ASTM D1125, USGS I-1780, USP 645, and for A2LA/NVLAP accreditations/ISO 9000 certifications. Material may be used to calibrate all conductivity meters and to determine all conductivity cell constants.

A2LA ACCREDITATION ISO/IEC 17025:2005 (1750.01)
• To assure accuracy an individually serial-numbered Traceable® Certificate is supplied to indicate traceability to standards provided by NIST (National Institute of Standards and Technology) and/or a National Standards Laboratory. A2LA accredited certification provides the highest achievable level of quality assurance, documentation, and accuracy. Conductivity Standards are produced in an A2LA accredited ISO/IEC 17025:2005 (1750.01) calibration laboratory. Additional certifications include ISO 31 (certificate content) and ISO 35 (statistical analysis), plus ISO 9001 (certified quality manufacturer).

Certificate of Analysis

Enter lot number
or click to view a Sample C of A

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat